Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACCGCACACAACGCACAATGAT[A/G]AAGAGCATAAACACAAGCCATAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610458 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZIC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LZIC - leucine zipper and CTNNBIP1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316973.1 | 800 | UTR 3 | NP_001303902.1 | |||
NM_001316974.1 | 800 | UTR 3 | NP_001303903.1 | |||
NM_001316975.1 | 800 | Intron | NP_001303904.1 | |||
NM_001316976.1 | 800 | UTR 3 | NP_001303905.1 | |||
NM_032368.4 | 800 | UTR 3 | NP_115744.2 | |||
XM_005263506.2 | 800 | Intron | XP_005263563.1 | |||
XM_005263509.2 | 800 | UTR 3 | XP_005263566.1 | |||
XM_017002546.1 | 800 | UTR 3 | XP_016858035.1 | |||
XM_017002547.1 | 800 | UTR 3 | XP_016858036.1 |