Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTTGTAAGTATTTTGTGCTTT[A/G]TTTTTGAGATATTTACTCACCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE4B - ubiquitination factor E4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105562.2 | Intron | NP_001099032.1 | ||||
NM_006048.4 | Intron | NP_006039.2 | ||||
XM_005263422.1 | Intron | XP_005263479.1 | ||||
XM_011540488.2 | Intron | XP_011538790.1 | ||||
XM_011540489.2 | Intron | XP_011538791.1 | ||||
XM_011540490.1 | Intron | XP_011538792.1 | ||||
XM_017000035.1 | Intron | XP_016855524.1 | ||||
XM_017000036.1 | Intron | XP_016855525.1 |