Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTAGTAGTTTTTGATTCCTGC[A/G]TAACAGTATTACCACAAATTTAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004565.2 | 85 | Intron | NP_004556.1 | |||
XM_005263470.4 | 85 | Intron | XP_005263527.1 | |||
XM_011541577.2 | 85 | Intron | XP_011539879.1 | |||
XM_011541578.2 | 85 | Intron | XP_011539880.1 | |||
XM_011541579.2 | 85 | UTR 5 | XP_011539881.1 | |||
XM_011541580.1 | 85 | Intron | XP_011539882.1 |