Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCAGCCCAAAGGGTACCTGTTC[C/T]TCCACTTGGCACTCTTGCCCTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609111 MIM: 605647 MIM: 604094 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO44 - F-box protein 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO6 - F-box protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018438.5 | Intron | NP_060908.1 | ||||
XM_005263448.4 | Intron | XP_005263505.1 | ||||
XM_005263449.4 | Intron | XP_005263506.1 | ||||
XM_005263451.4 | Intron | XP_005263508.1 | ||||
XM_006710571.3 | Intron | XP_006710634.1 | ||||
XM_011541232.2 | Intron | XP_011539534.1 | ||||
XM_017001007.1 | Intron | XP_016856496.1 |
MAD2L2 - MAD2 mitotic arrest deficient-like 2 (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |