Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGTGCTTTCCTTCGGGATGAGG[A/G]TTCACTGCACCTCCATTCCTAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGTRAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGTRAP - angiotensin II receptor associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040194.1 | Intron | NP_001035284.1 | ||||
NM_001040195.1 | Intron | NP_001035285.1 | ||||
NM_001040196.1 | Intron | NP_001035286.1 | ||||
NM_001040197.1 | Intron | NP_001035287.1 | ||||
NM_020350.4 | Intron | NP_065083.3 | ||||
XM_011541799.2 | Intron | XP_011540101.2 | ||||
XM_011541800.2 | Intron | XP_011540102.1 | ||||
XM_011541802.1 | Intron | XP_011540104.1 |