Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGTCTCTGTGCTGCTGCTGCA[A/G]GTGGTGCTGCTGCTGCGGTGAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602726 MIM: 607093 | ||||||||||||||||||||
Literature Links: |
CLCN6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCN6 - chloride voltage-gated channel 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256959.1 | 205 | Missense Mutation | AAG,AGG | K,R 13 | NP_001243888.1 | |
NM_001286.3 | 205 | Missense Mutation | AAG,AGG | K,R 13 | NP_001277.1 |
MTHFR - methylenetetrahydrofolate reductase (NAD(P)H) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005957.4 | 205 | Intron | NP_005948.3 | |||
XM_005263458.3 | 205 | Intron | XP_005263515.1 | |||
XM_005263460.4 | 205 | Intron | XP_005263517.1 | |||
XM_005263462.4 | 205 | Intron | XP_005263519.1 | |||
XM_005263463.3 | 205 | Intron | XP_005263520.1 | |||
XM_011541495.2 | 205 | Intron | XP_011539797.1 | |||
XM_011541496.2 | 205 | Intron | XP_011539798.1 | |||
XM_017001328.1 | 205 | Intron | XP_016856817.1 |