Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAACTGATTCTACTATTGAACCT[C/T]CTTTACATGGGATATTGGGTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608507 MIM: 153454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFN2 - mitofusin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127660.1 | Intron | NP_001121132.1 | ||||
NM_014874.3 | Intron | NP_055689.1 | ||||
XM_005263543.3 | Intron | XP_005263600.1 | ||||
XM_005263545.3 | Intron | XP_005263602.1 | ||||
XM_005263547.3 | Intron | XP_005263604.1 | ||||
XM_005263548.3 | Intron | XP_005263605.1 |
PLOD1 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |