Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCAGTCCCCTTTGTAGCAGGAA[A/G]AGCAGCTGGACACATGGGAAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFCP2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFCP2L1 - transcription factor CP2-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014553.2 | 7594 | UTR 3 | NP_055368.1 | |||
XM_017003902.1 | 7594 | UTR 3 | XP_016859391.1 | |||
XM_017003903.1 | 7594 | UTR 3 | XP_016859392.1 | |||
XM_017003904.1 | 7594 | Intron | XP_016859393.1 | |||
XM_017003905.1 | 7594 | Intron | XP_016859394.1 |