Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGCCATTGTGCTGCACACCTAC[C/G]TGGCAGTCATCCATCCACTGCGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR148 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR148 - G protein-coupled receptor 148 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207364.2 | 524 | Missense Mutation | CTG,GTG | L,V 148 | NP_997247.2 |
POTEI - POTE ankyrin domain family member I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277406.1 | 524 | Intron | NP_001264335.1 | |||
XM_011511670.2 | 524 | Intron | XP_011509972.1 | |||
XM_017004732.1 | 524 | Intron | XP_016860221.1 | |||
XM_017004733.1 | 524 | Intron | XP_016860222.1 | |||
XM_017004734.1 | 524 | Intron | XP_016860223.1 | |||
XM_017004735.1 | 524 | Intron | XP_016860224.1 |