Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGGAGGAGCTAGTCAATTAAA[A/G]CCAGACAGATTTTTGGGCTGTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604705 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MERTK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MERTK - MER proto-oncogene, tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006343.2 | Intron | NP_006334.2 | ||||
XM_005263565.4 | Intron | XP_005263622.1 | ||||
XM_005263568.4 | Intron | XP_005263625.1 | ||||
XM_011510490.2 | Intron | XP_011508792.1 | ||||
XM_017003164.1 | Intron | XP_016858653.1 | ||||
XM_017003165.1 | Intron | XP_016858654.1 |