Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATCTCATCCTACCCATGAGAAC[A/C]ATCATACTCCAGAGACTAAAAAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGAT5 - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM163 - transmembrane protein 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030923.4 | 1312 | UTR 3 | NP_112185.1 | |||
XM_011511950.2 | 1312 | UTR 3 | XP_011510252.1 | |||
XM_017005060.1 | 1312 | Intron | XP_016860549.1 | |||
XM_017005061.1 | 1312 | UTR 3 | XP_016860550.1 | |||
XM_017005062.1 | 1312 | Intron | XP_016860551.1 |