Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAGTAAACAAAGACAGTCTTAC[C/T]GACCTTTATGTCCAACATGCAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf49 - chromosome 2 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286537.1 | 481 | Silent Mutation | ACC,ACT | T,T 50 | NP_001273466.1 | |
NM_024093.2 | 481 | Silent Mutation | ACC,ACT | T,T 50 | NP_076998.1 | |
XM_017004892.1 | 481 | Silent Mutation | ACC,ACT | T,T 88 | XP_016860381.1 |
LOC100506473 - uncharacterized LOC100506473 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFBRAP1 - transforming growth factor beta receptor associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |