Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAGCAATGTGAGCTAACCAATG[C/T]GGTTGTTTTTAAGTGGCTGAGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1R2 - interleukin 1 receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261419.1 | Intron | NP_001248348.1 | ||||
NM_004633.3 | Intron | NP_004624.1 | ||||
XM_006712734.3 | Intron | XP_006712797.1 | ||||
XM_006712736.3 | Intron | XP_006712799.1 | ||||
XM_011511801.2 | Intron | XP_011510103.1 | ||||
XM_011511803.2 | Intron | XP_011510105.1 | ||||
XM_011511804.2 | Intron | XP_011510106.1 | ||||
XM_011511805.2 | Intron | XP_011510107.1 | ||||
XM_011511807.1 | Intron | XP_011510109.1 | ||||
XM_011511808.2 | Intron | XP_011510110.1 | ||||
XM_017004889.1 | Intron | XP_016860378.1 |