Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATTTAGAGAAAACTTCATGCC[G/T]TTGGGTGATTAGGTAAGTTTGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604494 MIM: 601203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL18R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL18R1 - interleukin 18 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282399.1 | Intron | NP_001269328.1 | ||||
NM_003855.3 | Intron | NP_003846.1 | ||||
XM_005264039.3 | Intron | XP_005264096.1 | ||||
XM_005264040.3 | Intron | XP_005264097.1 | ||||
XM_011512099.1 | Intron | XP_011510401.1 | ||||
XM_017005181.1 | Intron | XP_016860670.1 | ||||
XM_017005182.1 | Intron | XP_016860671.1 | ||||
XM_017005183.1 | Intron | XP_016860672.1 | ||||
XM_017005184.1 | Intron | XP_016860673.1 |
IL1RL1 - interleukin 1 receptor like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |