Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTCGAGGTAAAAGATTACGAC[A/G]GAGCATGGTTCTCTGTTCATGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATL2 - atlastin GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308076.1 | Intron | NP_001295005.1 | ||||
XM_011533017.1 | Intron | XP_011531319.1 | ||||
XM_011533018.2 | Intron | XP_011531320.1 | ||||
XM_011533019.2 | Intron | XP_011531321.1 | ||||
XM_011533020.1 | Intron | XP_011531322.1 | ||||
XM_011533022.2 | Intron | XP_011531324.1 | ||||
XM_011533023.2 | Intron | XP_011531325.1 | ||||
XM_017004686.1 | Intron | XP_016860175.1 | ||||
XM_017004687.1 | Intron | XP_016860176.1 | ||||
XM_017004688.1 | Intron | XP_016860177.1 |