Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGGGATGAAGGGAGGAAGCAC[A/C]ATTTAGAATGCTGTGCTATGTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EML4 - echinoderm microtubule associated protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145076.2 | Intron | NP_001138548.1 | ||||
NM_019063.4 | Intron | NP_061936.2 | ||||
XM_005264267.1 | Intron | XP_005264324.1 | ||||
XM_005264268.3 | Intron | XP_005264325.1 | ||||
XM_006711991.1 | Intron | XP_006712054.1 | ||||
XM_006711992.2 | Intron | XP_006712055.1 |
LOC102723824 - uncharacterized LOC102723824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |