Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCCAGACCTACCTCCTTATGG[T/A]CATGAGTGATGGCTGTGGAGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607788 MIM: 609332 | ||||||||||||||||||||
Literature Links: |
MCFD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCFD2 - multiple coagulation factor deficiency 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171506.2 | 437 | Intron | NP_001164977.1 | |||
NM_001171507.2 | 437 | Missense Mutation | GAC,GTC | D,V 100 | NP_001164978.1 | |
NM_001171508.2 | 437 | Missense Mutation | GAC,GTC | D,V 100 | NP_001164979.1 | |
NM_001171509.2 | 437 | Intron | NP_001164980.1 | |||
NM_001171510.2 | 437 | Intron | NP_001164981.1 | |||
NM_001171511.2 | 437 | Missense Mutation | GAC,GTC | D,V 81 | NP_001164982.1 | |
NM_139279.5 | 437 | Intron | NP_644808.1 |
TTC7A - tetratricopeptide repeat domain 7A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |