Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTAAAGCAACAGAAGACTTCTT[A/C]AGAAATAGCATCCAATCACCTCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605358 | ||||||||||||||||||||
Literature Links: |
GTF2A1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF2A1L - general transcription factor IIA subunit 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193487.2 | 2397 | Missense Mutation | TTA,TTC | L,F 23 | NP_001180416.1 | |
NM_006872.4 | 2397 | Missense Mutation | TTA,TTC | L,F 57 | NP_006863.2 |
STON1-GTF2A1L - STON1-GTF2A1L readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198593.1 | 2397 | Missense Mutation | TTA,TTC | L,F 761 | NP_001185522.1 | |
NM_001198594.1 | 2397 | Missense Mutation | TTA,TTC | L,F 761 | NP_001185523.1 | |
NM_172311.2 | 2397 | Missense Mutation | TTA,TTC | L,F 761 | NP_758515.1 |