Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGTTCAGAAAAAACACAAGAC[A/G]GGAAGGGGAAGAGGGGAAAAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC27 - DnaJ heat shock protein family (Hsp40) member C27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198559.1 | 2076 | UTR 3 | NP_001185488.1 | |||
NM_016544.2 | 2076 | UTR 3 | NP_057628.1 | |||
XM_011532902.1 | 2076 | Intron | XP_011531204.1 | |||
XM_017004269.1 | 2076 | UTR 3 | XP_016859758.1 | |||
XM_017004270.1 | 2076 | UTR 3 | XP_016859759.1 |