Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTAGTTCTCTTTTTAAGAGAGA[A/G]AGTGTGTTCCATTTACATTGGTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164910 | ||||||||||||||||||||
Literature Links: |
LINC01185 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01185 - long intergenic non-protein coding RNA 1185 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REL - REL proto-oncogene, NF-kB subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291746.1 | Intron | NP_001278675.1 | ||||
NM_002908.3 | Intron | NP_002899.1 | ||||
XM_011533010.2 | Intron | XP_011531312.1 | ||||
XM_017004627.1 | Intron | XP_016860116.1 |