Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAAGCCCAGCCTTACAAATACA[C/T]AAGAGTCAGAGAACCAAGTTTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERTAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERTAD2 - SERTA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014755.2 | 3883 | UTR 3 | NP_055570.1 | |||
XM_005264669.2 | 3883 | UTR 3 | XP_005264726.1 | |||
XM_011533204.1 | 3883 | UTR 3 | XP_011531506.1 | |||
XM_011533205.2 | 3883 | UTR 3 | XP_011531507.1 |