Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCTTTCTCTAATCTGAATATT[A/G]AGCTGAGCTGTGACTGGGTTAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616405 MIM: 106491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAK1 - AP2 associated kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ANXA4 - annexin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001153.4 | Intron | NP_001144.1 | ||||
NM_001320698.1 | Intron | NP_001307627.1 | ||||
NM_001320700.1 | Intron | NP_001307629.1 | ||||
NM_001320702.1 | Intron | NP_001307631.1 | ||||
XM_017003943.1 | Intron | XP_016859432.1 | ||||
XM_017003944.1 | Intron | XP_016859433.1 |