Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTTGGGCCATCACAAAGGAAG[A/G]TAAGGAGTATCCCCCTAGGAACCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF3C - kinesin family member 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002254.6 | 4166 | UTR 3 | NP_002245.4 | |||
XM_005264299.2 | 4166 | UTR 3 | XP_005264356.2 |