Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGACTGGAGCCAAGTGCGAGG[C/T]TGGCCTTTGCTGCAGAGGCCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602288 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf81 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf81 - chromosome 2 open reading frame 81 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTKN - rhotekin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015055.1 | 2398 | Silent Mutation | CAA,CAG | Q,Q 554 | NP_001015055.1 | |
NM_001015056.1 | 2398 | Silent Mutation | CAA,CAG | Q,Q 504 | NP_001015056.1 | |
NM_033046.2 | 2398 | Silent Mutation | CAA,CAG | Q,Q 541 | NP_149035.1 | |
XM_005264478.2 | 2398 | Intron | XP_005264535.1 | |||
XM_005264479.4 | 2398 | Intron | XP_005264536.1 | |||
XM_017004635.1 | 2398 | Silent Mutation | CAA,CAG | Q,Q 541 | XP_016860124.1 | |
XM_017004636.1 | 2398 | UTR 3 | XP_016860125.1 | |||
XM_017004637.1 | 2398 | UTR 3 | XP_016860126.1 | |||
XM_017004638.1 | 2398 | Silent Mutation | CAA,CAG | Q,Q 424 | XP_016860127.1 | |
XM_017004639.1 | 2398 | Intron | XP_016860128.1 | |||
XM_017004640.1 | 2398 | Intron | XP_016860129.1 |
WDR54 - WD repeat domain 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320823.1 | 2398 | Intron | NP_001307752.1 | |||
NM_001320824.1 | 2398 | Intron | NP_001307753.1 | |||
NM_001320825.1 | 2398 | Intron | NP_001307754.1 | |||
NM_032118.3 | 2398 | Intron | NP_115494.1 | |||
XM_017005064.1 | 2398 | Intron | XP_016860553.1 |