Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTATTCCGCACTTTGATATCTGT[C/T]GATTATTTCTGGAGCAAAGGCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ADGRF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRF3 - adhesion G protein-coupled receptor F3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145168.1 | 3744 | Intron | NP_001138640.1 | |||
NM_001145169.1 | 3744 | UTR 3 | NP_001138641.1 | |||
NM_001321971.1 | 3744 | UTR 3 | NP_001308900.1 | |||
NM_001321975.1 | 3744 | UTR 3 | NP_001308904.1 | |||
NM_153835.3 | 3744 | UTR 3 | NP_722577.2 | |||
XM_011532621.2 | 3744 | Intron | XP_011530923.1 | |||
XM_011532622.2 | 3744 | Intron | XP_011530924.1 | |||
XM_011532623.2 | 3744 | Intron | XP_011530925.1 | |||
XM_017003505.1 | 3744 | Intron | XP_016858994.1 | |||
XM_017003506.1 | 3744 | UTR 3 | XP_016858995.1 |