Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTCAATTTTTCTGTTACCACT[A/G]TTTGGTGCATAGGATCCATTTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRF3 - adhesion G protein-coupled receptor F3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145168.1 | Intron | NP_001138640.1 | ||||
NM_001145169.1 | Intron | NP_001138641.1 | ||||
NM_001321971.1 | Intron | NP_001308900.1 | ||||
NM_001321975.1 | Intron | NP_001308904.1 | ||||
NM_153835.3 | Intron | NP_722577.2 | ||||
XM_011532621.2 | Intron | XP_011530923.1 | ||||
XM_011532622.2 | Intron | XP_011530924.1 | ||||
XM_011532623.2 | Intron | XP_011530925.1 | ||||
XM_017003505.1 | Intron | XP_016858994.1 | ||||
XM_017003506.1 | Intron | XP_016858995.1 |