Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCTACAGATCCAGTGCATTCAA[A/G]ATGCCAACCAGATTTTGCTCACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186910 MIM: 186730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD8A - CD8a molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD8B - CD8b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178100.1 | Intron | NP_001171571.1 | ||||
NM_004931.4 | Intron | NP_004922.1 | ||||
NM_172101.3 | Intron | NP_742099.1 | ||||
NM_172102.3 | Intron | NP_742100.1 | ||||
NM_172213.3 | Intron | NP_757362.1 | ||||
XM_011533164.2 | Intron | XP_011531466.1 |