Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGACCTCAGATGAAGAGCTGGTC[C/T]GAAATACAGAATTGGGAATTATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf16 - chromosome 2 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271286.1 | Intron | NP_001258215.1 | ||||
NM_001271287.1 | Intron | NP_001258216.1 | ||||
NM_001271288.1 | Intron | NP_001258217.1 | ||||
NM_001271289.1 | Intron | NP_001258218.1 | ||||
NM_001271318.1 | Intron | NP_001258247.1 | ||||
NM_032434.3 | Intron | NP_115810.2 |