Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTTTTATTGGAATGGGGTGAG[C/T]TTGGAGAAAAGAAGGCATTTCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610497 MIM: 611132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRE - brain and reproductive organ-expressed (TNFRSF1A modulator) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261840.1 | Intron | NP_001248769.1 | ||||
NM_004899.4 | Intron | NP_004890.2 | ||||
NM_199191.2 | Intron | NP_954661.1 | ||||
NM_199192.2 | Intron | NP_954662.1 | ||||
NM_199193.2 | Intron | NP_954663.1 | ||||
NM_199194.2 | Intron | NP_954664.1 |
BRE-AS1 - BRE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBKS - ribokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |