Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCTCGTTGTACTTGTCCTTGAG[T/C]ACGTAGCCCAGCTCCCGAAACTGTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601570 | |||||||||||||||||||||||
Literature Links: |
WNT7A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
WNT7A - Wnt family member 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004625.3 | 986 | Silent Mutation | GTA,GTG | V,V 227 | NP_004616.2 | |
XM_011534091.2 | 986 | Silent Mutation | GTA,GTG | V,V 160 | XP_011532393.1 |