Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAGCAAAACTTTTAAAACAAAC[A/G]AGATAAACTCACTTCTTTCCCCAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605652 MIM: 609784 | |||||||||||||||||||||||
Literature Links: |
FBXL2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FBXL2 - F-box and leucine rich repeat protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171713.1 | 3846 | Intron | NP_001165184.1 | |||
NM_012157.3 | 3846 | Intron | NP_036289.3 | |||
XM_005265014.2 | 3846 | Intron | XP_005265071.1 | |||
XM_005265015.3 | 3846 | Intron | XP_005265072.1 | |||
XM_005265016.2 | 3846 | Intron | XP_005265073.1 | |||
XM_005265017.3 | 3846 | Intron | XP_005265074.1 | |||
XM_011533559.2 | 3846 | Intron | XP_011531861.1 | |||
XM_017006092.1 | 3846 | Intron | XP_016861581.1 | |||
XM_017006093.1 | 3846 | Intron | XP_016861582.1 | |||
XM_017006094.1 | 3846 | Intron | XP_016861583.1 |
UBP1 - upstream binding protein 1 (LBP-1a) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128160.1 | 3846 | UTR 3 | NP_001121632.1 | |||
NM_001128161.1 | 3846 | UTR 3 | NP_001121633.1 | |||
NM_014517.4 | 3846 | UTR 3 | NP_055332.3 |