Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTCTGCGCCCCTCCTCGCTGCG[A/C]ACCCAAGATATCTCGCACTTGCTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604050 MIM: 602142 | |||||||||||||||||||||||
Literature Links: |
DLEC1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DLEC1 - deleted in lung and esophageal cancer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321153.1 | 894 | Silent Mutation | CGA,CGC | R,R 88 | NP_001308082.1 | |
NM_007335.3 | 894 | Silent Mutation | CGA,CGC | R,R 88 | NP_031361.2 | |
NM_007337.3 | 894 | Silent Mutation | CGA,CGC | R,R 88 | NP_031363.2 | |
XM_006713438.3 | 894 | Silent Mutation | CGA,CGC | R,R 88 | XP_006713501.1 | |
XM_006713439.3 | 894 | Silent Mutation | CGA,CGC | R,R 88 | XP_006713502.1 | |
XM_006713440.3 | 894 | Silent Mutation | CGA,CGC | R,R 88 | XP_006713503.1 | |
XM_011534317.2 | 894 | Silent Mutation | CGA,CGC | R,R 88 | XP_011532619.1 | |
XM_011534318.2 | 894 | Silent Mutation | CGA,CGC | R,R 88 | XP_011532620.1 |
PLCD1 - phospholipase C delta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |