Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGAGTTACATACTTTCTTTTTT[C/T]CCCAGTGGATACCCTCACATTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 192321 | ||||||||||||||||||||
Literature Links: |
VIPR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VIPR1 - vasoactive intestinal peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251882.1 | Intron | NP_001238811.1 | ||||
NM_001251883.1 | Intron | NP_001238812.1 | ||||
NM_001251884.1 | Intron | NP_001238813.1 | ||||
NM_001251885.1 | Intron | NP_001238814.1 | ||||
NM_004624.3 | Intron | NP_004615.2 | ||||
XM_005265437.2 | Intron | XP_005265494.1 | ||||
XM_005265438.3 | Intron | XP_005265495.1 | ||||
XM_005265439.2 | Intron | XP_005265496.1 | ||||
XM_011534079.1 | Intron | XP_011532381.1 | ||||
XM_011534080.2 | Intron | XP_011532382.1 |