Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTGTTTTCTTTGCCCCAGTTT[G/T]TCTGTGATATCAACTGGAGCAATAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604780 MIM: 613726 | |||||||||||||||||||||||
Literature Links: |
ABHD5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ABHD5 - abhydrolase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016006.4 | Intron | NP_057090.2 | ||||
XM_011533779.2 | Intron | XP_011532081.1 | ||||
XM_011533780.1 | Intron | XP_011532082.1 |
ANO10 - anoctamin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |