Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAAAAGATATCTTCAACCCA[A/G]CACGCATTTCAGTGAGTCAGAACTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 164975 | |||||||||||||||||||||||
Literature Links: |
WNT5A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
WNT5A - Wnt family member 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256105.1 | 3036 | UTR 3 | NP_001243034.1 | |||
NM_003392.4 | 3036 | UTR 3 | NP_003383.2 | |||
XM_006713324.1 | 3036 | UTR 3 | XP_006713387.1 | |||
XM_011534085.2 | 3036 | UTR 3 | XP_011532387.1 | |||
XM_011534086.2 | 3036 | UTR 3 | XP_011532388.1 | |||
XM_011534087.2 | 3036 | UTR 3 | XP_011532389.1 | |||
XM_011534088.2 | 3036 | UTR 3 | XP_011532390.1 | |||
XM_011534089.1 | 3036 | UTR 3 | XP_011532391.1 | |||
XM_017007127.1 | 3036 | UTR 3 | XP_016862616.1 | |||
XM_017007128.1 | 3036 | UTR 3 | XP_016862617.1 |