Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCATGCTCCTCAGGGATTGGCTC[C/T]GGCCAAGTGCCCCAGCCTCCTTTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612115 MIM: 609869 | |||||||||||||||||||||||
Literature Links: |
ARHGEF3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARHGEF3 - Rho guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128615.1 | 1167 | Intron | NP_001122087.1 | |||
NM_001128616.1 | 1167 | Intron | NP_001122088.1 | |||
NM_001289698.1 | 1167 | Intron | NP_001276627.1 | |||
NM_019555.2 | 1167 | Intron | NP_062455.1 | |||
XM_005265186.4 | 1167 | Intron | XP_005265243.1 | |||
XM_005265187.3 | 1167 | Intron | XP_005265244.1 | |||
XM_011533764.1 | 1167 | Intron | XP_011532066.1 | |||
XM_011533765.1 | 1167 | Intron | XP_011532067.1 | |||
XM_011533766.2 | 1167 | Intron | XP_011532068.1 | |||
XM_011533767.1 | 1167 | Intron | XP_011532069.1 | |||
XM_017006502.1 | 1167 | Intron | XP_016861991.1 | |||
XM_017006503.1 | 1167 | Intron | XP_016861992.1 | |||
XM_017006504.1 | 1167 | Intron | XP_016861993.1 | |||
XM_017006505.1 | 1167 | Intron | XP_016861994.1 | |||
XM_017006506.1 | 1167 | Intron | XP_016861995.1 |
SPATA12 - spermatogenesis associated 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181727.1 | 1167 | UTR 5 | NP_859078.1 | |||
XM_011533676.2 | 1167 | UTR 5 | XP_011531978.1 |