Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATATAATATCCATGAAATCAAT[A/C]TGCTTCAATGCAGTGTGAAAGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUFY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUFY3 - RUN and FYVE domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037442.3 | Intron | NP_001032519.1 | ||||
NM_001130709.1 | Intron | NP_001124181.1 | ||||
NM_001291993.1 | Intron | NP_001278922.1 | ||||
NM_001291994.1 | Intron | NP_001278923.1 | ||||
NM_014961.4 | Intron | NP_055776.1 | ||||
XM_005265656.2 | Intron | XP_005265713.1 | ||||
XM_005265658.1 | Intron | XP_005265715.1 | ||||
XM_011531750.2 | Intron | XP_011530052.1 | ||||
XM_011531751.2 | Intron | XP_011530053.1 | ||||
XM_011531752.2 | Intron | XP_011530054.1 | ||||
XM_011531753.2 | Intron | XP_011530055.1 | ||||
XM_011531754.2 | Intron | XP_011530056.1 | ||||
XM_011531755.2 | Intron | XP_011530057.1 | ||||
XM_011531756.2 | Intron | XP_011530058.1 | ||||
XM_017007890.1 | Intron | XP_016863379.1 | ||||
XM_017007891.1 | Intron | XP_016863380.1 | ||||
XM_017007892.1 | Intron | XP_016863381.1 |