Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGCAAGCTTTCGGAAACAAAA[C/T]AGGCAACATTTTGTGATCATAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601903 | ||||||||||||||||||||
Literature Links: |
UGT2B17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT2B17 - UDP glucuronosyltransferase family 2 member B17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077.3 | 1585 | Silent Mutation | CTA,CTG | L,L 514 | NP_001068.1 |