Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTGCCTTTCTGATTCCCTGGAG[G/T]TCATTTCATAGCTGAGCTTCTTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601851 MIM: 614726 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLOCK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLOCK - clock circadian regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267843.1 | 2858 | UTR 3 | NP_001254772.1 | |||
NM_004898.3 | 2858 | UTR 3 | NP_004889.1 | |||
XM_005265787.2 | 2858 | UTR 3 | XP_005265844.1 | |||
XM_011534409.2 | 2858 | UTR 3 | XP_011532711.1 | |||
XM_011534410.2 | 2858 | UTR 3 | XP_011532712.1 | |||
XM_011534411.2 | 2858 | UTR 3 | XP_011532713.1 | |||
XM_017008854.1 | 2858 | UTR 3 | XP_016864343.1 | |||
XM_017008855.1 | 2858 | UTR 3 | XP_016864344.1 |
TMEM165 - transmembrane protein 165 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018475.4 | 2858 | Intron | NP_060945.2 | |||
XM_011534394.2 | 2858 | Intron | XP_011532696.1 | |||
XM_017008412.1 | 2858 | Intron | XP_016863901.1 |