Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGTTGTCAAGATCTTACATAC[C/T]TTTACACTGGGGCGGCAGTAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOPX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOPX - HOP homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145459.1 | 858 | Intron | NP_001138931.1 | |||
NM_001145460.1 | 858 | Missense Mutation | AAA,GAA | K,E 110 | NP_001138932.1 | |
NM_032495.5 | 858 | Intron | NP_115884.4 | |||
NM_139211.4 | 858 | Intron | NP_631957.1 | |||
NM_139212.3 | 858 | Intron | NP_631958.1 | |||
XM_006714052.2 | 858 | Missense Mutation | AGT,GGT | S,G 110 | XP_006714115.1 | |
XM_017008728.1 | 858 | Intron | XP_016864217.1 | |||
XM_017008729.1 | 858 | Intron | XP_016864218.1 | |||
XM_017008730.1 | 858 | Intron | XP_016864219.1 | |||
XM_017008731.1 | 858 | Intron | XP_016864220.1 | |||
XM_017008732.1 | 858 | Intron | XP_016864221.1 | |||
XM_017008733.1 | 858 | Intron | XP_016864222.1 | |||
XM_017008734.1 | 858 | Intron | XP_016864223.1 |