Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACAGACTTAAAGAGGCACAACT[C/T]AGCATCACCCTACTTAGCTTGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TENM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TENM2 - teneurin transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080428.2 | Intron | NP_001073897.2 | ||||
NM_001122679.1 | Intron | NP_001116151.1 | ||||
XM_005265950.2 | Intron | XP_005266007.1 | ||||
XM_005265952.2 | Intron | XP_005266009.1 | ||||
XM_006714897.2 | Intron | XP_006714960.1 | ||||
XM_011534604.2 | Intron | XP_011532906.1 | ||||
XM_017009660.1 | Intron | XP_016865149.1 | ||||
XM_017009661.1 | Intron | XP_016865150.1 | ||||
XM_017009662.1 | Intron | XP_016865151.1 | ||||
XM_017009663.1 | Intron | XP_016865152.1 | ||||
XM_017009664.1 | Intron | XP_016865153.1 | ||||
XM_017009665.1 | Intron | XP_016865154.1 | ||||
XM_017009666.1 | Intron | XP_016865155.1 | ||||
XM_017009667.1 | Intron | XP_016865156.1 | ||||
XM_017009668.1 | Intron | XP_016865157.1 | ||||
XM_017009669.1 | Intron | XP_016865158.1 |