Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCCTATGGAAGTGGCATATTTT[C/G]ATCAGAAACCTGTTCTGTATGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK2 - dedicator of cytokinesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004946.2 | Intron | NP_004937.1 | ||||
XM_005265830.3 | Intron | XP_005265887.1 | ||||
XM_011534448.2 | Intron | XP_011532750.1 | ||||
XM_011534449.2 | Intron | XP_011532751.1 | ||||
XM_011534450.2 | Intron | XP_011532752.1 | ||||
XM_011534451.2 | Intron | XP_011532753.1 | ||||
XM_017009189.1 | Intron | XP_016864678.1 | ||||
XM_017009190.1 | Intron | XP_016864679.1 |