Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCGCAGGACGGCTACGGTACG[C/G]GGGTGGGAGGGCTTCGGAGCACGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3912 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3912 - microRNA 3912 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPM1 - nucleophosmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037738.2 | Intron | NP_001032827.1 | ||||
NM_002520.6 | Intron | NP_002511.1 | ||||
NM_199185.3 | Intron | NP_954654.1 | ||||
XM_005265920.3 | Intron | XP_005265977.1 | ||||
XM_011534564.1 | Intron | XP_011532866.1 |