Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTTCACTGTTGCATCGGTTG[A/G]TAAAGTCAATCAGCATTGCTTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREBRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREBRF - CREB3 regulatory factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168393.1 | Intron | NP_001161865.1 | ||||
NM_001168394.1 | Intron | NP_001161866.1 | ||||
NM_153607.2 | Intron | NP_705835.2 | ||||
XM_005265821.3 | Intron | XP_005265878.1 | ||||
XM_006714822.3 | Intron | XP_006714885.1 | ||||
XM_011534442.1 | Intron | XP_011532744.1 |