Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAGCTTTCAAAAGGTGTCTGGC[A/G]TATTCTAAAAAGGAAAGATAATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616745 | ||||||||||||||||||||
Literature Links: |
BOD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BOD1 - biorientation of chromosomes in cell division 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159651.1 | 883 | Missense Mutation | CGC,TGC | R,C 123 | NP_001153123.1 | |
NM_138369.2 | 883 | UTR 3 | NP_612378.1 | |||
XM_017010017.1 | 883 | Missense Mutation | ACG,ATG | T,M 170 | XP_016865506.1 |