Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAAGGCATGTGGGCGGCAGCTAA[C/T]GCAGAATTTTCTGAGCCACACGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600869 | ||||||||||||||||||||
Literature Links: |
GRK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRK6 - G protein-coupled receptor kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004105.2 | 477 | Missense Mutation | ACG,ATG | T,M 106 | NP_001004105.1 | |
NM_001004106.2 | 477 | Missense Mutation | ACG,ATG | T,M 106 | NP_001004106.1 | |
NM_002082.3 | 477 | Missense Mutation | ACG,ATG | T,M 106 | NP_002073.2 | |
XM_006714858.3 | 477 | Missense Mutation | ACG,ATG | T,M 76 | XP_006714921.1 | |
XM_006714859.2 | 477 | Missense Mutation | ACG,ATG | T,M 74 | XP_006714922.1 | |
XM_011534537.2 | 477 | Missense Mutation | ACG,ATG | T,M 67 | XP_011532839.1 | |
XM_011534538.1 | 477 | Intron | XP_011532840.1 |
PRR7-AS1 - PRR7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |