Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGATTTAAGGAAGTTACTGAA[C/T]CTCTGCTATTAGGCCTATCATAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604147 MIM: 605974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTTG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTTG1 - pituitary tumor-transforming 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282382.1 | Intron | NP_001269311.1 | ||||
NM_001282383.1 | Intron | NP_001269312.1 | ||||
NM_004219.3 | Intron | NP_004210.1 |
SLU7 - SLU7 homolog, splicing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |