Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAAGGAAACAAGGAATCAAGAA[A/C]ACATACATTATTCACAGGTGGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRB2 - gamma-aminobutyric acid type A receptor beta2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000813.2 | 3477 | UTR 3 | NP_000804.1 | |||
NM_021911.2 | 3477 | UTR 3 | NP_068711.1 |