Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGGGGGGCTGCTACTTTCTTA[C/G]AACTAGAAGCTGAGCAATGTATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRA1 - gamma-aminobutyric acid type A receptor alpha1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000806.5 | Intron | NP_000797.2 | ||||
NM_001127643.1 | Intron | NP_001121115.1 | ||||
NM_001127644.1 | Intron | NP_001121116.1 | ||||
NM_001127645.1 | Intron | NP_001121117.1 | ||||
NM_001127648.1 | Intron | NP_001121120.1 |