Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAACGTGGTGCCCAGGACAGG[C/T]GGATCCAGGATCTGGAAACTGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMMR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMMR - hyaluronan mediated motility receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142556.1 | 492 | Missense Mutation | CGG,TGG | R,W 105 | NP_001136028.1 | |
NM_001142557.1 | 492 | Missense Mutation | CGG,TGG | R,W 18 | NP_001136029.1 | |
NM_012484.2 | 492 | Missense Mutation | CGG,TGG | R,W 104 | NP_036616.2 | |
NM_012485.2 | 492 | Missense Mutation | CGG,TGG | R,W 89 | NP_036617.2 |
NUDCD2 - NudC domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |